1-(3-Bromophenyl)-2-thiourea

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-(3-Bromophenyl)thiourea, 3-Bromophenylthiourea, (3-Bromophenyl)thiourea, N-(3-Bromophenyl)thiourea

OT-0934-5G OT-0934-1G OT-0934-25G
COBBOT-0934-5GEA 0 GBP
COBBOT-0934-5G COBBOT-0934-1G COBBOT-0934-25G
1-(3-Bromophenyl)-2-thiourea
1-(3-Bromophenyl)-2-thiourea
Formula: (C₆H₄Br)NHC(S)NH₂
MW: 231.12 g/mol
MDL Number: MFCD00060440
CAS Number: 21327-14-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR