1-(3-Bromophenyl)-2-thiourea

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-(3-Bromophenyl)thiourea, 3-Bromophenylthiourea, (3-Bromophenyl)thiourea, N-(3-Bromophenyl)thiourea

AS24976G25 AS24976G5
ACTIAS24976G25EA 0 GBP
ACTIAS24976G25 ACTIAS24976G5
1-(3-Bromophenyl)-2-thiourea
1-(3-Bromophenyl)-2-thiourea
Formula: (C₆H₄Br)NHC(S)NH₂
MW: 231.12 g/mol
MDL Number: MFCD00060440
CAS Number: 21327-14-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR