1-(2-Bromophenyl)-2-thiourea
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-(2-Bromophenyl)thiourea, 2-Bromophenylthiourea, 1-(2-Bromophenyl)thiourea
OT-0917-5G
OT-0917-1G
OT-0917-25G
COBBOT-0917-5GEA
0
GBP
COBBOT-0917-5G
COBBOT-0917-1G
COBBOT-0917-25G
1-(2-Bromophenyl)-2-thiourea
1-(2-Bromophenyl)-2-thiourea
Formula:
(C₆H₄Br)NHC(S)NH₂ MW: 231.12 g/mol Boiling Pt: 319 °C (1013 hPa) Melting Pt: 130…132.5 °C |
MDL Number:
MFCD00041146 CAS Number: 5391-30-0 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...