3'-Fluoropropiophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one

AS24936G100 AS24936G25 AS24936G5
ACTIAS24936G100EA 0 GBP
ACTIAS24936G100 ACTIAS24936G25 ACTIAS24936G5
3'-Fluoropropiophenone
3'-Fluoropropiophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
MDL Number: MFCD00061245
CAS Number: 455-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR