3',5'-Dihydroxyacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone

AS24581G100 AS24581G25
ACTIAS24581G100EA 99 GBP
ACTIAS24581G100 ACTIAS24581G25
3',5'-Dihydroxyacetophenone
3',5'-Dihydroxyacetophenone
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00002290
CAS Number: 51863-60-6
EINECS: 257-480-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR