Benzophenone, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

M-Clarity™ quality level = MQ100

73664-5G-F
73664-5G-FEA 54.9 GBP
73664-5G-F
Benzophenone, Supelco®
Benzophenone
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305.4
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9 III
Merck Index: 13,01099
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13132c96fea4aa6ed0715e0b15a57e42dcbd86a5&vid=4f80c9e21bad451d48d48c4aecfb573f70e9190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR