4-Chloroindole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Chloro-1H-indole
016227-1G
016227-5G
FLUO016227-1GEA
0
GBP
FLUO016227-1G
FLUO016227-5G
4-Chloroindole
4-Chloroindole
Formula:
C₈H₆ClN MW: 151.6 g/mol Boiling Pt: 293 Melting Pt: 84…86 °C Density: 1.259 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005665 CAS Number: 25235-85-2 EINECS: 246-747-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...