4'-tert-Butyl-2-chloroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(tert-Butyl)phenacyl chloride
011112-5G
011112-25G
FLUO011112-5GEA
0
GBP
FLUO011112-5G
FLUO011112-25G
4'-tert-Butyl-2-chloroacetophenone
4'-tert-Butyl-2-chloroacetophenone
Formula:
C₉H₉ClO MW: 210.71 g/mol |
MDL Number:
MFCD00171621 CAS Number: 21886-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...