1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

AS12378G1 AS12378G5
ACTIAS12378G1EA 16 GBP
ACTIAS12378G1 ACTIAS12378G5
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₄H₂₃BN₂O₃
MW: 278.1550 g/mol
MDL Number: MFCD16556148
CAS Number: 1003846-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR