6-(Trifluoromethyl)nicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008113-1G 008113-5G 008113-10G
FLUO008113-1GEA 0 GBP
FLUO008113-1G FLUO008113-5G FLUO008113-10G
6-(Trifluoromethyl)nicotinic acid
6-(Trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
MDL Number: MFCD00792430
CAS Number: 231291-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR