2-Piperazinone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Piperazin-2-one
M-Clarity™ quality level = MQ100
641065-1G
641065-5G
SIAL641065-1GEA
38.8
GBP
SIAL641065-1G
SIAL641065-5G
2-Piperazinone, Sigma-Aldrich®
2-Piperazinone
Formula:
C₄H₈N₂O MW: 100.12 g/mol |
MDL Number:
MFCD01318687 CAS Number: 5625-67-2 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...