4-Methoxy-3-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxy-3-methylbenzeneboronic acid

BB-2157-5G BB-2157-25G BB-2157-1G
COBBBB-2157-5GEA 0 GBP
COBBBB-2157-5G COBBBB-2157-25G COBBBB-2157-1G
4-Methoxy-3-methylphenylboronic acid
4-Methoxy-3-methylphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
MDL Number: MFCD02179464
CAS Number: 175883-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR