2-(p-Tolyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24523G100 AS24523G25
ACTIAS24523G100EA 0 GBP
ACTIAS24523G100 ACTIAS24523G25
2-(p-Tolyl)pyridine
2-(p-Tolyl)pyridine
Formula: C₁₂H₁₁N
MW: 169.22 g/mol
MDL Number: MFCD00006281
CAS Number: 4467-06-5
EINECS: 224-734-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR