2-Chloro-5-fluoroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20818G1 AS20818G5
ACTIAS20818G1EA 0 GBP
ACTIAS20818G1 ACTIAS20818G5
2-Chloro-5-fluoroisonicotinic acid
2-Chloro-5-fluoroisonicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175.5500 g/mol
MDL Number: MFCD04972381
CAS Number: 884494-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR