2-Amino-3,4-dimethylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-3,4-dimethylbenzoic Acid, ['3,4-Dimethylanthranilic acid']

AS22268G100 AS22268G25
ACTIAS22268G100EA 109 GBP
ACTIAS22268G100 ACTIAS22268G25
2-Amino-3,4-dimethylbenzoic acid
2-Amino-3,4-dimethylbenzoic acid
Formula: C₉H₁₁NO₂
MDL Number: MFCD00130041
CAS Number: 50419-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR