4-Chloro-2-ethoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-2-ethoxybenzeneboronic acid

BB-2477-5G BB-2477-25G BB-2477-1G
COBBBB-2477-5GEA 0 GBP
COBBBB-2477-5G COBBBB-2477-25G COBBBB-2477-1G
4-Chloro-2-ethoxyphenylboronic acid
4-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MDL Number: MFCD03094940
CAS Number: 850568-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR