3-Chloro-4-ethoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Chloro-4-ethoxyphenyl)boronic acid

BB-2725-5G BB-2725-25G BB-2725-1G
COBBBB-2725-5GEA 0 GBP
COBBBB-2725-5G COBBBB-2725-25G COBBBB-2725-1G
3-Chloro-4-ethoxyphenylboronic acid
3-Chloro-4-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200.4300 g/mol
MDL Number: MFCD03701556
CAS Number: 279261-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR