2-Butylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-8488-5G
BB-8488-1G
COBBBB-8488-5GEA
0
GBP
COBBBB-8488-5G
COBBBB-8488-1G
2-Butylboronic acid
2-Butylboronic acid
Formula:
C₄H₁₁BO₂ |
MDL Number:
MFCD06656265 CAS Number: 88496-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...