2-Chloro-m-anisidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-methoxyaniline

AS31002G1 AS31002G5
ACTIAS31002G1EA 0 GBP
ACTIAS31002G1 ACTIAS31002G5
2-Chloro-m-anisidine
2-Chloro-m-anisidine
Formula: C₇H₈ClNO
MW: 157.6000 g/mol
MDL Number: MFCD08691737
CAS Number: 113206-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR