5-Amino-2-chlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23519G1 AS23519G5
ACTIAS23519G1EA 0 GBP
ACTIAS23519G1 ACTIAS23519G5
5-Amino-2-chlorobenzonitrile
5-Amino-2-chlorobenzonitrile
Formula: C₇H₅ClN₂
MDL Number: MFCD00800459
CAS Number: 35747-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR