3-Amino-5-chlorobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4347-5G CA-4347-1G
COBBCA-4347-5GEA 0 GBP
COBBCA-4347-5G COBBCA-4347-1G
3-Amino-5-chlorobenzonitrile
3-Amino-5-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD06797224
CAS Number: 53312-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR