4-Amino-3-chlorobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

519596-25G 519596-5G
SIAL519596-25GEA 658 GBP
SIAL519596-25G SIAL519596-5G
4-Amino-3-chlorobenzonitrile, Sigma-Aldrich®
4-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00052917
CAS Number: 21803-75-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR