5-Bromo-2-chlorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS31197G1 AS31197G5
ACTIAS31197G1EA 14 GBP
ACTIAS31197G1 ACTIAS31197G5
5-Bromo-2-chlorobenzaldehyde
5-Bromo-2-chlorobenzaldehyde
Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD08445659
CAS Number: 189628-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR