2,2,3,4,4,4-Hexafluorobutyl methacrylate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2,3,4,4,4-Hexafluorobutyl Methacrylate
002400-5G
002400-25G
002400-100G
FLUO002400-5GEA
0
GBP
FLUO002400-5G
FLUO002400-25G
FLUO002400-100G
2,2,3,4,4,4-Hexafluorobutyl methacrylate
2,2,3,4,4,4-Hexafluorobutyl methacrylate
Formula:
C₈H₈F₆O₂ |
MDL Number:
MFCD00042311 CAS Number: 36405-47-7 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...