3-Bromo-5-chloro-2-methylaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21878G1 AS21878G5
ACTIAS21878G1EA 0 GBP
ACTIAS21878G1 ACTIAS21878G5
3-Bromo-5-chloro-2-methylaniline
3-Bromo-5-chloro-2-methylaniline
Formula: C₇H₇BrClN
MW: 220.4942 g/mol
MDL Number: MFCD12198860
CAS Number: 1166756-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR