2-Fluoro-3-(trifluoromethyl)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006441-1G 006441-5G
FLUO006441-1GEA 0 GBP
FLUO006441-1G FLUO006441-5G
2-Fluoro-3-(trifluoromethyl)phenylacetonitrile
2-Fluoro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
MDL Number: MFCD00061178
CAS Number: 239087-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR