3-Fluoro-4-(trifluoromethyl)benzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007915-1G 007915-5G 007915-10G
FLUO007915-1GEA 0 GBP
FLUO007915-1G FLUO007915-5G FLUO007915-10G
3-Fluoro-4-(trifluoromethyl)benzophenone
3-Fluoro-4-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
MDL Number: MFCD00236282
CAS Number: 243128-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR