2-Fluoro-6-(trifluoromethyl)benzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006398-1G 006398-5G 006398.10G
FLUO006398-1GEA 0 GBP
FLUO006398-1G FLUO006398-5G FLUO006398.10G
2-Fluoro-6-(trifluoromethyl)benzophenone
2-Fluoro-6-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
MDL Number: MFCD00061255
CAS Number: 208173-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR