1-(5-Fluoro-2-methylphenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(5-Fluoro-2-methylphenyl)thiourea

008348-1G 008348-5G
FLUO008348-1GEA 0 GBP
FLUO008348-1G FLUO008348-5G
1-(5-Fluoro-2-methylphenyl)-2-thiourea
1-(5-Fluoro-2-methylphenyl)-2-thiourea
Formula: C₈H₉FN₂S
MW: 184.23 g/mol
MDL Number: MFCD00052380
CAS Number: 16822-86-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR