1-(4-Isopropylphenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018757-1G 018757-5G
FLUO018757-1GEA 0 GBP
FLUO018757-1G FLUO018757-5G
1-(4-Isopropylphenyl)-2-thiourea
1-(4-Methylphenyl)-2-thiourea
Formula: C₁₀H₁₄N₂S
MDL Number: MFCD00060459
CAS Number: 65259-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR