5-Chloro-2-nitro-p-toluidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS20806G25 AS20806G5
ACTIAS20806G25EA 0 GBP
ACTIAS20806G25 ACTIAS20806G5
5-Chloro-2-nitro-p-toluidine
5-Chloro-2-nitro-p-toluidine
Formula: C₇H₇ClN₂O₂
MDL Number: MFCD00034066
CAS Number: 7149-80-6
UN: 2237
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR