5-Bromo-2-methyl-3-nitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24871G1 AS24871G5
ACTIAS24871G1EA 0 GBP
ACTIAS24871G1 ACTIAS24871G5
5-Bromo-2-methyl-3-nitroaniline
5-Bromo-2-methyl-3-nitroaniline
Formula: C₇H₇BrN₂O₂
MW: 231.0500 g/mol
MDL Number: MFCD07781362
CAS Number: 864550-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR