6-Nitro-o-anisidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Nitro-o-anisidine, 2-Amino-3-nitroanisole, 2-Methoxy-6-nitrophenylamine, 2-Methoxy-6-nitroaniline

AS21648G1 AS21648G25 AS21648G5
ACTIAS21648G1EA 0 GBP
ACTIAS21648G1 ACTIAS21648G25 ACTIAS21648G5
6-Nitro-o-anisidine
6-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
MDL Number: MFCD01930197
CAS Number: 16554-45-3
EINECS: 240-617-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR