Hexyl chloroformate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

252778-5G 252778-25G
SIAL252778-5GEA 73.6 GBP
SIAL252778-5G SIAL252778-25G
Hexyl chloroformate, Sigma-Aldrich®
Hexyl chloroformate
Formula: C₇H₁₃ClO₂
MDL Number: MFCD00000651
CAS Number: 6092-54-2
EINECS: 228-036-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR