5-Fluoronicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4058-5G
CA-4058-25G
CA-4058-1G
COBBCA-4058-5GEA
0
GBP
COBBCA-4058-5G
COBBCA-4058-25G
COBBCA-4058-1G
5-Fluoronicotinic acid
5-Fluoronicotinic acid
Formula:
C₆H₄FNO₂ MW: 141.10 g/mol |
MDL Number:
MFCD01318537 CAS Number: 402-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...