5-Fluoronicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21780G100 AS21780G25 AS21780G5
ACTIAS21780G100EA 0 GBP
ACTIAS21780G100 ACTIAS21780G25 ACTIAS21780G5
5-Fluoronicotinic acid
5-Fluoronicotinic acid
Formula: C₆H₄FNO₂
MW: 141.10 g/mol
MDL Number: MFCD01318537
CAS Number: 402-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR