2,2,2-Trifluoroethyl methanesulphonate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008580-5G
008580-100G
008580-25G
FLUO008580-5GEA
0
GBP
FLUO008580-5G
FLUO008580-100G
FLUO008580-25G
2,2,2-Trifluoroethyl methanesulphonate
2,2,2-Trifluoroethyl methanesulphonate
Formula:
C₃H₅F₃O₃S MW: 178.130 g/mol |
MDL Number:
MFCD00025058 CAS Number: 25236-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...