Ethyl pentafluoropropionate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001175-25G 001175-100G
FLUO001175-25GEA 0 GBP
FLUO001175-25G FLUO001175-100G
Ethyl pentafluoropropionate
Ethyl pentafluoropropionate
Formula: C₅H₅F₅O₂
MDL Number: MFCD00000431
CAS Number: 426-65-3
EINECS: 207-043-6
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR