2'-Bromo-5'-chloroacetanilide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4703-5G
CA-4703-25G
CA-4703-1G
COBBCA-4703-5GEA
0
GBP
COBBCA-4703-5G
COBBCA-4703-25G
COBBCA-4703-1G
2'-Bromo-5'-chloroacetanilide
2'-Bromo-5'-chloroacetanilide
Formula:
C₈H₇BrClNO |
MDL Number:
MFCD12026007 CAS Number: 827-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...