Potassium bis(trimethylsilyl)amide 0.5 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
277304-100ML
277304-800ML
SIAL277304-100MLEA
162
GBP
SIAL277304-100ML
SIAL277304-800ML
Potassium bis(trimethylsilyl)amide 0.5 mol/l in toluene, Sigma-Aldrich®
Potassium bis(trimethylsilyl)amide
Formula:
C₆H₁₈KNSi₂ |
MDL Number:
MFCD00010330 CAS Number: 40949-94-8 EINECS: 424-100-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...