2-Isopropoxyaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22227G1 AS22227G25 AS22227G5
ACTIAS22227G1EA 0 GBP
ACTIAS22227G1 ACTIAS22227G25 ACTIAS22227G5
2-Isopropoxyaniline
2-Isopropoxyaniline
Formula: C₉H₁₃NO
MDL Number: MFCD00242947
CAS Number: 29026-74-2
EINECS: 249-377-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR