4-Nitro-2,5-xylidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dimethyl-4-nitroaniline

AS21395G1 AS21395G5
ACTIAS21395G1EA 0 GBP
ACTIAS21395G1 ACTIAS21395G5
4-Nitro-2,5-xylidine
4-Nitro-2,5-xylidine
Formula: C₈H₁₀N₂O₂
MW: 166.1772 g/mol
MDL Number: MFCD00025207
CAS Number: 3460-29-5
EINECS: 222-405-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR