4-Amino-2-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22181G1 AS22181G5
ACTIAS22181G1EA 0 GBP
ACTIAS22181G1 ACTIAS22181G5
4-Amino-2-(trifluoromethyl)pyridine
4-Amino-2-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.1100 g/mol
MDL Number: MFCD09056877
CAS Number: 147149-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR