6-Fluoro-o-toluic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Fluoro-6-methylbenzoic acid
AS21590G1
AS21590G25
AS21590G5
ACTIAS21590G1EA
0
GBP
ACTIAS21590G1
ACTIAS21590G25
ACTIAS21590G5
6-Fluoro-o-toluic acid
6-Fluoro-o-toluic acid
Formula:
C₈H₇FO₂ MW: 154.1400 g/mol |
MDL Number:
MFCD06658162 CAS Number: 90259-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...