2-Fluoro-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-3-methylbenzoic acid

AS24118G25 AS24118G5
ACTIAS24118G25EA 0 GBP
ACTIAS24118G25 ACTIAS24118G5
2-Fluoro-m-toluic acid
2-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00042504
CAS Number: 315-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR