5-Fluoro-2-methylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20646G100 AS20646G25 AS20646G5
ACTIAS20646G100EA 0 GBP
ACTIAS20646G100 ACTIAS20646G25 ACTIAS20646G5
5-Fluoro-2-methylbenzaldehyde
5-Fluoro-2-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
MDL Number: MFCD02261763
CAS Number: 22062-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR