3-Bromo-o-cresol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromo-2-methylphenol
AS34356G1
AS34356G5
ACTIAS34356G1EA
0
GBP
ACTIAS34356G1
ACTIAS34356G5
3-Bromo-o-cresol
3-Bromo-o-cresol
Formula:
C₇H₇BrO MW: 187.0339 g/mol |
MDL Number:
MFCD11100990 CAS Number: 7766-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...