3'-Bromo-4'-morpholinoacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21562G25 AS21562G5
ACTIAS21562G25EA 0 GBP
ACTIAS21562G25 ACTIAS21562G5
3'-Bromo-4'-morpholinoacetophenone
3'-Bromo-4'-morpholinoacetophenone
Formula: C₁₂H₁₄BrNO₂
MDL Number: MFCD09878364
CAS Number: 957066-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR