3-Thiopheneboronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Thiophene-3-boronic acid pinacol ester, Thiophene-3-boronic acid, pinacol ester

021636-5G 021636-1G
FLUO021636-5GEA 0 GBP
FLUO021636-5G FLUO021636-1G
3-Thiopheneboronic acid pinacol ester
3-Thiopheneboronic acid pinacol ester
Formula: C₁₀H₁₅BO₂S
MW: 210.1009 g/mol
MDL Number: MFCD05663893
CAS Number: 214360-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR