6-(2-Fluorophenyl)-3-pyridinecarbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24486G1 AS24486G5
ACTIAS24486G1EA 0 GBP
ACTIAS24486G1 ACTIAS24486G5
6-(2-Fluorophenyl)-3-pyridinecarbaldehyde
6-(2-Fluorophenyl)-3-pyridinecarbaldehyde
Formula: C₁₂H₈FNO
MDL Number: MFCD04115443
CAS Number: 898795-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR